Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-hydroxy-3-iodobenzoate, 98%

Chemical Identifiers

Chemical Name or Material Methyl 4-hydroxy-3-iodobenzoate
CAS 15126-06-4
Formula Weight 278.04
MDL Number MFCD05664378
Synonym 4-Hydroxy-3-iodobenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5687706

  SDS
alfa aesar™
H5687706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 155° to 159°C
Formula Weight 278.04
Color White to pale brown
Physical Form Crystal or Powder
Chemical Name or Material Methyl 4-hydroxy-3-iodobenzoate
Health Hazard 2 GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Synonym 4-Hydroxy-3-iodobenzoic acid methyl ester
Sensitivity Light Sensitive
CAS 15126-06-4
MDL Number MFCD05664378
Quantity 5g
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P310-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Assay 98%
Safety and Handling

Danger

GHS P Statement
P280-P264-P305+P351+P338-P310-P301+P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.