missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4'-formylbiphenyl-4-carboxylate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444780010

 View more versions of this product

Catalog No. AC444780010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Methyl 4'-formylbiphenyl-4-carboxylate
113.0°C to 118.0°C
70916-89-1
C15H12O3
URINKTWOCVLGGR-UHFFFAOYSA-N
methyl 4-(4-formylphenyl)benzoate
4321323
97%
Glass Bottle
1g
96% min. (GC)
1,1'-biphenyl-4,4'-dicarboxylicacid, 4-methyl ester, 1,1'-biphenyl-4-carboxylicacid,4'-formyl-,methyl ester, 4'-formylbiphenyl-4-carboxylic acid methyl ester, acmc-209ogh, methyl 4'-formyl 1,1'-biphenyl-4-carboxylate, methyl 4'-formyl-1,1'-biphenyl-4-carboxylate, methyl 4'-formylbiphenyl-4-carboxylate, methyl 4-4-formylphenyl benzoate, methyl-4'-formylbiphenyl-4-carboxylate
COC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C=O
240.26
240.26

Chemical Identifiers

70916-89-1
240.26
1,1'-biphenyl-4,4'-dicarboxylicacid, 4-methyl ester, 1,1'-biphenyl-4-carboxylicacid,4'-formyl-,methyl ester, 4'-formylbiphenyl-4-carboxylic acid methyl ester, acmc-209ogh, methyl 4'-formyl 1,1'-biphenyl-4-carboxylate, methyl 4'-formyl-1,1'-biphenyl-4-carboxylate, methyl 4'-formylbiphenyl-4-carboxylate, methyl 4-4-formylphenyl benzoate, methyl-4'-formylbiphenyl-4-carboxylate
methyl 4-(4-formylphenyl)benzoate
C15H12O3
URINKTWOCVLGGR-UHFFFAOYSA-N
4321323
COC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C=O
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents