Please login to your online account to display your discounted pricing

Methyl 4-formylbenzoate, 99%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  223750100

 View more versions of this product

Catalog No. AC223750100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder or Chunks
Assay ≥98.5%
Formula Weight 164.16
Quantity 10g
Color White to cream or slightly green
EINECS Number 216-385-5
TSCA TSCA
Melting Point 59° to 63°C
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 99%
Linear Formula HCOC6H4CO2CH3
Synonym Methyl terephthalaldehydate
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Methyl 4-formylbenzoate
Name Note 99%
Molecular Formula C9H8O3
MDL Number MFCD00006950
CAS 1571-08-0

Specifications

Chemical Name or Material Methyl 4-formylbenzoate
Name Note 99%
CAS 1571-08-0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H8O3
Linear Formula HCOC6H4CO2CH3
Formula Weight 164.16
MDL Number MFCD00006950
Synonym Methyl terephthalaldehydate
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents