Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-formyl-2,5-dimethylpyrrole-3-carboxylate, 96%

$167.26 - $742.84

Chemical Identifiers

Chemical Name or Material Methyl 4-formyl-2,5-dimethylpyrrole-3-carboxylate
CAS 175205-91-1
Molecular Formula C9H11NO3
Formula Weight 181.19
MDL Number MFCD00203870
Synonym 4-Formyl-2,5-dimethyl-1H-pyrrole-3-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5047703

  SDS
alfa aesar™
H5047703
1g Each for $167.26

AAH5047706

  SDS
alfa aesar™
H5047706
5g Each for $742.84
Description & Specifications

Specifications

Formula Weight 181.19
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Methyl 4-formyl-2,5-dimethylpyrrole-3-carboxylate
Synonym 4-Formyl-2,5-dimethyl-1H-pyrrole-3-carboxylic acid methyl ester
Quantity 1g
Molecular Formula C9H11NO3
Assay 96%
MDL Number MFCD00203870
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 175205-91-1
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.