Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-fluorosalicylate, 98+%

$91.46 - $266.70

Chemical Identifiers

Chemical Name or Material Methyl 4-fluorosalicylate
CAS 392-04-1
Molecular Formula C8H7FO3
Formula Weight 170.14
MDL Number MFCD02093965
Beilstein 2087920
Synonym 4-Fluorosalicylic acid methyl ester; Methyl 4-fluoro-2-hydroxybenzoate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1784603

  SDS
alfa aesar™
L1784603
1g Each for $266.70

AAL17846MD

  SDS
alfa aesar™
L17846MD
250mg Each for $91.46
Description & Specifications

Specifications

Formula Weight 170.14
Chemical Name or Material Methyl 4-fluorosalicylate
Assay ≥98%
Molecular Formula C8H7FO3
MDL Number MFCD02093965
CAS 392-04-1
Beilstein 2087920
Synonym 4-Fluorosalicylic acid methyl ester; Methyl 4-fluoro-2-hydroxybenzoate
Health Hazard 1 Warning
Melting Point 38° to 40°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.