Please login to your online account to display your discounted pricing

Methyl 4-fluorophenylacetate, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  304670250

 View more versions of this product

Catalog No. AC304670250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (GC)
Formula Weight 168.06
Physical Form Liquid
Quantity 25g
Color Clear colorless to yellow
CAS 34837-84-8
Assay Percent Range 99%
Linear Formula FC6H4CH2CO2CH3
Chemical Name or Material Methyl 4-fluorophenylacetate
Refractive Index 1.4860 to 1.4880 (20°C, 589nm)
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C9H9FO2
Health Hazard 2 GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material Methyl 4-fluorophenylacetate
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C9H9FO2
Linear Formula FC6H4CH2CO2CH3
Formula Weight 168.06
Accessories

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents