missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 4-fluorobenzoylacetate, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2554603
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl 4-fluorobenzoylacetate | |
1.228 | |
>110°C (230°F) | |
1.521 | |
1 g | |
HGLVYXXPRSNKQN-UHFFFAOYSA-N | |
methyl 3-(4-fluorophenyl)-3-oxopropanoate | |
579425 | |
95% |
63131-29-3 | |
91°C (5 mmHg) | |
C10H9FO3 | |
MFCD00000355 | |
methyl 4-fluorobenzoylacetate, methyl 3-4-fluorophenyl-3-oxopropanoate, methyl p-fluorobenzoylacetate, 4-fluorobenzoylacetic acid methyl ester, methyl 4'-fluorobenzoylacetate, 3-4-fluoro-phenyl-3-oxo-propionic acid methyl ester, benzenepropanoic acid, 4-fluoro-beta-oxo-, methyl ester, benzenepropanoic acid, 4-fluoro-.beta.-oxo-, methyl ester, pubchem16721, acmc-209nd6 | |
COC(=O)CC(=O)C1=CC=C(C=C1)F | |
196.177 | |
196.18 |
Chemical Identifiers
63131-29-3 | |
196.177 | |
HGLVYXXPRSNKQN-UHFFFAOYSA-N | |
579425 | |
COC(=O)CC(=O)C1=CC=C(C=C1)F |
C10H9FO3 | |
MFCD00000355 | |
methyl 4-fluorobenzoylacetate, methyl 3-4-fluorophenyl-3-oxopropanoate, methyl p-fluorobenzoylacetate, 4-fluorobenzoylacetic acid methyl ester, methyl 4'-fluorobenzoylacetate, 3-4-fluoro-phenyl-3-oxo-propionic acid methyl ester, benzenepropanoic acid, 4-fluoro-beta-oxo-, methyl ester, benzenepropanoic acid, 4-fluoro-.beta.-oxo-, methyl ester, pubchem16721, acmc-209nd6 | |
methyl 3-(4-fluorophenyl)-3-oxopropanoate |
Safety and Handling
EINECSNumber : 263-889-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only