Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-fluoro-3-hydroxybenzoate, 98+%

$88.15 - $256.15

Chemical Identifiers

Chemical Name or Material Methyl 4-fluoro-3-hydroxybenzoate
CAS 214822-96-5
Molecular Formula C8H7FO3
Formula Weight 170.14
MDL Number MFCD06797457
Synonym 4-Fluoro-3-hydroxybenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2568306

  SDS
alfa aesar™
B2568306
5g Each for $88.15

AAB2568314

  SDS
alfa aesar™
B2568314
25g Each for $256.15
Description & Specifications

Specifications

Formula Weight 170.14
MDL Number MFCD06797457
Synonym 4-Fluoro-3-hydroxybenzoic acid methyl ester
Health Hazard 1 Warning
Chemical Name or Material Methyl 4-fluoro-3-hydroxybenzoate
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H7FO3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 214822-96-5
Assay ≥98%
Melting Point 89° to 91°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.