Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-fluoro-2-methylbenzoate, 98%

Chemical Identifiers

Chemical Name or Material Methyl 4-fluoro-2-methylbenzoate
CAS 174403-69-1
Formula Weight 168.16
MDL Number MFCD06203790
Synonym 4-Fluoro-2-methylbenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6326806

  SDS
alfa aesar™
H6326806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6326814

  SDS
alfa aesar™
H6326814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6326822

  SDS
alfa aesar™
H6326822
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 168.16
MDL Number MFCD06203790
Assay 98%
Physical Form Liquid
CAS 174403-69-1
Color Colorless
Chemical Name or Material Methyl 4-fluoro-2-methylbenzoate
Synonym 4-Fluoro-2-methylbenzoic acid methyl ester
Quantity 5g