missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 4-ethynylbenzoate, 97%, Thermo Scientific Chemicals
$247.14 - $918.78
Chemical Identifiers
CAS | 3034-86-4 |
---|---|
Molecular Formula | C10H8O2 |
Molecular Weight (g/mol) | 160.17 |
MDL Number | MFCD00168820 |
InChI Key | JPGRSTBIEYGVNO-UHFFFAOYSA-N |
Synonym | 4-ethynylbenzoic acid methyl ester, 4-ethynyl-benzoic acid methyl ester, benzoic acid, 4-ethynyl-, methyl ester, methyl4-ethynylbenzoate, 4-ethynylbenzoic acid methylester, 4-methoxycarbonyl phenylacetylene, benzoic acid,4-ethynyl-, methyl ester, methyl 4-ethynyl-benzoate, 4-ethinylbenzoic acid methyl ester, 4-ethynyl benzoic acid methylester |
PubChem CID | 640163 |
IUPAC Name | methyl 4-ethynylbenzoate |
SMILES | COC(=O)C1=CC=C(C=C1)C#C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5341903
|
Thermo Scientific Chemicals
H5341903 |
1 g |
Each for $247.14
|
|
|||||
AAH5341906
|
Thermo Scientific Chemicals
H5341906 |
5 g |
Each for $918.78
|
|
|||||
Description
Methyl 4-ethynylbenzoate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 4-ethynylbenzoate is used as pharmaceutical intermediate.
Solubility
Sparingly soluble in water (0.16 g/L at 25°C).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
3034-86-4 | |
160.17 | |
JPGRSTBIEYGVNO-UHFFFAOYSA-N | |
640163 | |
COC(=O)C1=CC=C(C=C1)C#C |
C10H8O2 | |
MFCD00168820 | |
4-ethynylbenzoic acid methyl ester, 4-ethynyl-benzoic acid methyl ester, benzoic acid, 4-ethynyl-, methyl ester, methyl4-ethynylbenzoate, 4-ethynylbenzoic acid methylester, 4-methoxycarbonyl phenylacetylene, benzoic acid,4-ethynyl-, methyl ester, methyl 4-ethynyl-benzoate, 4-ethinylbenzoic acid methyl ester, 4-ethynyl benzoic acid methylester | |
methyl 4-ethynylbenzoate |
Specifications
3034-86-4 | |
C10H8O2 | |
1 g | |
Sparingly soluble in water (0.16g/L at 25°C). | |
COC(=O)C1=CC=C(C=C1)C#C | |
160.17 | |
160.17 | |
Methyl 4-ethynylbenzoate |
92°C to 94°C | |
MFCD00168820 | |
4-ethynylbenzoic acid methyl ester, 4-ethynyl-benzoic acid methyl ester, benzoic acid, 4-ethynyl-, methyl ester, methyl4-ethynylbenzoate, 4-ethynylbenzoic acid methylester, 4-methoxycarbonyl phenylacetylene, benzoic acid,4-ethynyl-, methyl ester, methyl 4-ethynyl-benzoate, 4-ethinylbenzoic acid methyl ester, 4-ethynyl benzoic acid methylester | |
JPGRSTBIEYGVNO-UHFFFAOYSA-N | |
methyl 4-ethynylbenzoate | |
640163 | |
97% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only