Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-ethoxybenzoate, 98%

$884.47 - $884.47

Chemical Identifiers

Chemical Name or Material Methyl 4-ethoxybenzoate
CAS 23676-08-6
Molecular Formula C10H12O3
Formula Weight 180.21
MDL Number MFCD00017264
Beilstein 2691838
Synonym 4-Ethoxybenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1798022

  SDS
alfa aesar™
A1798022
100g Each for $884.47
Description & Specifications

Specifications

Formula Weight 180.21
Chemical Name or Material Methyl 4-ethoxybenzoate
Assay 98%
Molecular Formula C10H12O3
MDL Number MFCD00017264
CAS 23676-08-6
Beilstein 2691838
Synonym 4-Ethoxybenzoic acid methyl ester
Melting Point 35° to 37°C
Boiling Point 160°C (20mmHg)
Quantity 100g
Flash Point >110°C (230°F)

Applications
Methyl 4-ethoxybenzoate is used in the preparation of 4- ethoxybenzoic acid by reacting with aluminum tribromide. It is involved in the preparation of apricoxib, which is a selective cyclooxygenase inhibitor.

Solubility
Soluble in alcohol and ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.