Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-diethylaminobenzoate, 98%

Chemical Identifiers

Chemical Name or Material Methyl 4-diethylaminobenzoate
CAS 91563-80-3
Molecular Formula C12H17NO2
Formula Weight 207.26
MDL Number MFCD00189408
Synonym 4-Diethylaminobenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1738418

 
alfa aesar™
A1738418
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Methyl 4-diethylaminobenzoate
CAS 91563-80-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
Molecular Formula C12H17NO2
Formula Weight 207.26
MDL Number MFCD00189408
Synonym 4-Diethylaminobenzoic acid methyl ester
Melting Point 46° to 47°C
Quantity 50g