missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-chlorophenylacetate, 99%, ACROS Organics™

Chemical Identifiers

CAS 52449-43-1
Assay Percent Range 99%
Molecular Formula C9H9ClO2
Linear Formula ClC6H4CHCOCH3
Formula Weight 184.62
MDL Number MFCD00032743
Chemical Name or Material Methyl 4-chlorophenylacetate, 99%
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC351050050 SDS Fisher Scientific Acros
35105-0050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC351050250 SDS Fisher Scientific Acros
35105-0250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

52449-43-1
C9H9ClO2
184.62
Methyl 4-chlorophenylacetate, 99%
99%
ClC6H4CHCOCH3
MFCD00032743
≥98.5%

Specifications

1.2g/mL
5g
Authentic
1.5220 to 1.5240 (20°C, 589nm)
99%
ClC6H4CHCOCH3
MFCD00032743
Methyl 4-chlorophenylacetate, 99%
Clear slightly yellow
52449-43-1
Glass bottle
Liquid
C9H9ClO2
184.62
09, III, 2264
≥98.5%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 257-927-0