Learn More
Methyl 4-chlorophenylacetate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 351050050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyl 4-chlorophenylacetate | |
52449-43-1 | |
Authentic | |
Glass bottle | |
1.5220 to 1.5240 | |
MFCD00032743 | |
09, III, 2264 | |
methyl 4-chlorophenylacetate, methyl 2-4-chlorophenyl acetate, methyl 4-chlorophenyl acetate, 4-chloro-phenyl-acetic acid methyl ester, benzeneacetic acid, 4-chloro-, methyl ester, 4-chlorophenylacetic acid methyl ester, methyl4-chlorophenylacetate, zlchem 382 | |
COC(=O)CC1=CC=C(Cl)C=C1 | |
184.62 | |
184.62 |
99% | |
1.2000g/mL | |
98.5% min. (GC) | |
C9H9ClO2 | |
ClC6H4CH2CO2CH3 | |
5g | |
1.2 | |
WWIYGBWRUXQDND-UHFFFAOYSA-N | |
methyl 2-(4-chlorophenyl)acetate | |
104196 | |
99% |
Chemical Identifiers
52449-43-1 | |
184.62 | |
WWIYGBWRUXQDND-UHFFFAOYSA-N | |
104196 | |
COC(=O)CC1=CC=C(Cl)C=C1 |
C9H9ClO2 | |
MFCD00032743 | |
methyl 4-chlorophenylacetate, methyl 2-4-chlorophenyl acetate, methyl 4-chlorophenyl acetate, 4-chloro-phenyl-acetic acid methyl ester, benzeneacetic acid, 4-chloro-, methyl ester, 4-chlorophenylacetic acid methyl ester, methyl4-chlorophenylacetate, zlchem 382 | |
methyl 2-(4-chlorophenyl)acetate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 257-927-0
RUO â Research Use Only