missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl (4-Chlorophenyl)acetate 98.0+%, TCI America™

$44.44 - $110.28

Chemical Identifiers

CAS 52449-43-1
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00032743
InChI Key WWIYGBWRUXQDND-UHFFFAOYSA-N
Synonym 4-chloro-phenyl-acetic acid methyl ester, 4-chlorophenylacetic acid methyl ester, benzeneacetic acid, 4-chloro-, methyl ester, methyl 2-4-chlorophenyl acetate, methyl 4-chlorophenyl acetate, methyl 4-chlorophenylacetate, methyl4-chlorophenylacetate, zlchem 382
PubChem CID 104196
IUPAC Name methyl 2-(4-chlorophenyl)acetate
SMILES COC(=O)CC1=CC=C(C=C1)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M20395G View Documents Tci America
M20395G
5g Each for $44.44
 
M203925G View Documents Tci America
M203925G
25g Each for $110.28
 
Specifications

Chemical Identifiers

52449-43-1
184.619
WWIYGBWRUXQDND-UHFFFAOYSA-N
104196
COC(=O)CC1=CC=C(C=C1)Cl
C9H9ClO2
MFCD00032743
4-chloro-phenyl-acetic acid methyl ester, 4-chlorophenylacetic acid methyl ester, benzeneacetic acid, 4-chloro-, methyl ester, methyl 2-4-chlorophenyl acetate, methyl 4-chlorophenyl acetate, methyl 4-chlorophenylacetate, methyl4-chlorophenylacetate, zlchem 382
methyl 2-(4-chlorophenyl)acetate

Specifications

115°C
5g
C9H9ClO2
4-chloro-phenyl-acetic acid methyl ester, 4-chlorophenylacetic acid methyl ester, benzeneacetic acid, 4-chloro-, methyl ester, methyl 2-4-chlorophenyl acetate, methyl 4-chlorophenyl acetate, methyl 4-chlorophenylacetate, methyl4-chlorophenylacetate, zlchem 382
COC(=O)CC1=CC=C(C=C1)Cl
184.619
184.62
≥98.0% (GC)
Yellow
52449-43-1
MFCD00032743
WWIYGBWRUXQDND-UHFFFAOYSA-N
methyl 2-(4-chlorophenyl)acetate
104196
Clear Liquid at 20°C
Methyl (4-Chlorophenyl)acetate
Safety and Handling

Safety and Handling

TSCA : No