Please login to your online account to display your discounted pricing

Methyl 4-(chloromethyl)benzoate 97% , ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC391310050

 View more versions of this product

Catalog No. AC391310050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 184.62
Physical Form Low Melting Solid
Quantity 5g
Color White
Melting Point 37° to 40°C
Linear Formula ClCH2C6H4C(O)OCH3
CAS 34040-64-7
Infrared Spectrum Authentic
Chemical Name or Material Methyl 4-(chloromethyl)benzoate
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00511224
Packaging Glass bottle
Molecular Formula C9H9ClO2
Assay Percent Range 97%
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

Specifications

Chemical Name or Material Methyl 4-(chloromethyl)benzoate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H9ClO2
Linear Formula ClCH2C6H4C(O)OCH3
Formula Weight 184.62
MDL Number MFCD00511224
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents