Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-(chloromethyl)benzoate, 97%

Chemical Identifiers

Chemical Name or Material Methyl 4-(chloromethyl)benzoate
CAS 34040-64-7
Molecular Formula C9H9ClO2
Formula Weight 184.62
MDL Number MFCD00511224
UN Number UN3261
Synonym 4-(Chloromethyl)benzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2247604

  SDS
alfa aesar™
B2247604
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2247609

  SDS
alfa aesar™
B2247609
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 184.62
Quantity 2g
Melting Point 39° to 40°C
CAS 34040-64-7
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Assay 97%
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 4-(Chloromethyl)benzoic acid methyl ester
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Chemical Name or Material Methyl 4-(chloromethyl)benzoate
Boiling Point 160° to 165°C (30mmHg)
Molecular Formula C9H9ClO2
UN Number UN3261
MDL Number MFCD00511224
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.