Please login to your online account to display your discounted pricing

Methyl 4-(chloroformyl)butyrate 98%, ACROS Organics™

$210.14 - $210.14

Chemical Identifiers

Chemical Name or Material Methyl 4-(chloroformyl)butyrate
Name Note 98%
CAS 1501-26-4
Assay Percent Range 98%
Molecular Formula C6H9ClO3
Linear Formula CO(CH2)3CO2CH3
Formula Weight 164.59
MDL Number MFCD00000756
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC126540250

  SDS
thermo fisher scientific
126540250
Glass bottle 25mL Each for $210.14

AC126540100

  SDS
thermo fisher scientific
AC126540100
Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 164.59
Quantity 25mL
Density 1.1910g/mL
EINECS Number 216-115-6
Chemical Name or Material Methyl 4-(chloroformyl)butyrate
Name Note 98%
Assay Percent Range 98%
Molecular Formula C6H9ClO3
Linear Formula ClCO(CH2)3CO2CH3
MDL Number MFCD00000756
CAS 1501-26-4
Synonym Methyl glutaryl chloride
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protectio
Packaging Glass bottle
Safety and Handling

Danger

GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protectio

GHS P Statement Causes severe skin burns and eye damage.