Please login to your online account to display your discounted pricing

Methyl 4-(chloroformyl)butyrate 98%, ACROS Organics™


Quantity 25mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  126540250

 View more versions of this product

Catalog No. AC126540250

  • / Each
 

Description & Specifications

Chemical Identifiers

Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician.
Formula Weight 164.59
Quantity 25mL
Density 1.1910g/mL
EINECS Number 216-115-6
Health Hazard 1 Danger
Packaging Glass bottle
Synonym Methyl glutaryl chloride
Linear Formula ClCO(CH2)3CO2CH3
Molecular Formula C6H9ClO3
Assay Percent Range 98%
Chemical Name or Material Methyl 4-(chloroformyl)butyrate
Name Note 98%
CAS 1501-26-4
MDL Number MFCD00000756

Specifications

Chemical Name or Material Methyl 4-(chloroformyl)butyrate
Name Note 98%
CAS 1501-26-4
Assay Percent Range 98%
Molecular Formula C6H9ClO3
Linear Formula ClCO(CH2)3CO2CH3
Formula Weight 164.59
MDL Number MFCD00000756
Synonym Methyl glutaryl chloride
Accessories

Danger

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
Causes severe skin burns and eye damage.

SDS & Safety Documents