missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-chlorocarbonylbenzoate, 98+%

Manufacturer:  Alfa Aesar™ H5468803

 View more versions of this product

Catalog No. AAH5468803



Description

Description

Applications
4-(Chlorocarbonyl)benzoic Acid Methyl Ester, is a building block, that can be used for the synthesis of various pharmaceutical compounds. It can be used for the preparation of benzophenone residues and also for the synthesis of novel novel bulky hydrophobic retinoids.

Solubility
Hydrolyzes in water.

Notes
Store at room temperature. Keep away from oxidizing agents and acids. Hygroscopic.
Specifications

Specifications

Methyl 4-chlorocarbonylbenzoate
52°C to 56°C
7377-26-6
MFCD00059486
4-carbomethoxybenzoylchloride
CVXXHXPNTZBZEL-UHFFFAOYSA-N
methyl 4-carbonochloridoylbenzoate
81850
≥98%
Moisture sensitive
1g
C9H7ClO3
UN3261
Hydrolyzes in water.
COC(=O)C1=CC=C(C=C1)C(=O)Cl
198.602
198.6

Chemical Identifiers

7377-26-6
198.602
CVXXHXPNTZBZEL-UHFFFAOYSA-N
81850
COC(=O)C1=CC=C(C=C1)C(=O)Cl
C9H7ClO3
MFCD00059486
4-carbomethoxybenzoylchloride
methyl 4-carbonochloridoylbenzoate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 230-937-2

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

Documents