missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-chlorobutyrate, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC126520050

 View more versions of this product

Catalog No. AC126520050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Liquid
136.58
Clear colorless
Glass Bottle
1.4311 to 1.4331 (20°C, 589nm)
C5H9ClO2
98+%
3153-37-5
≥98.0%
5g
1.1200g/mL
Authentic
Cl(CH2)3CO2CH3
Methyl 4-chlorobutyrate
98+%
MFCD00001003

Chemical Identifiers

3153-37-5
C5H9ClO2
136.58
Methyl 4-chlorobutyrate, 98+%
98+%
Cl(CH2)3CO2CH3
MFCD00001003
≥98.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 221-592-9

TSCA : TSCA