Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4'-chlorobiphenyl-4-carboxylate, 95%

$55.45 - $219.99

Chemical Identifiers

Chemical Name or Material Methyl 4'-chlorobiphenyl-4-carboxylate
CAS 89901-02-0
Molecular Formula C14H11ClO2
Formula Weight 246.69
MDL Number MFCD04117794
Synonym 4'-Chlorobiphenyl-4-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5191003

  SDS
alfa aesar™
H5191003
1g Each for $55.45

AAH5191006

  SDS
alfa aesar™
H5191006
5g Each for $219.99
Description & Specifications

Specifications

Formula Weight 246.69
Quantity 1g
Molecular Formula C14H11ClO2
MDL Number MFCD04117794
Synonym 4'-Chlorobiphenyl-4-carboxylic acid methyl ester
Assay 95%
CAS 89901-02-0
Chemical Name or Material Methyl 4'-chlorobiphenyl-4-carboxylate