Learn More
Methyl 4-chloroacetoacetate, 97+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6470914
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl 4-chloroacetoacetate | |
Colorless | |
85°C (4 mmHg) | |
C5H7ClO3 | |
MFCD00000938 | |
UN2922 | |
methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042 | |
COC(=O)CC(=O)CCl | |
150.56 | |
150.56 | |
Liquid |
32807-28-6 | |
1.305 g/mL | |
102°C (216°F) | |
1.456 | |
25 g | |
1759939 | |
HFLMYYLFSNEOOT-UHFFFAOYSA-N | |
methyl 4-chloro-3-oxobutanoate | |
36240 | |
≥97% |
Chemical Identifiers
32807-28-6 | |
150.56 | |
HFLMYYLFSNEOOT-UHFFFAOYSA-N | |
36240 | |
COC(=O)CC(=O)CCl |
C5H7ClO3 | |
MFCD00000938 | |
methyl 4-chloroacetoacetate, methyl 4-chloro-3-oxo-butanoate, butanoic acid, 4-chloro-3-oxo-, methyl ester, 4-chloroacetoacetic acid methyl ester, methyl 4-chloro-3-oxobutyrate, methyl gamma-chloroacetoacetate, methyl-4-chloroacetoacetate, acetoacetic acid, 4-chloro-, methyl ester, 4-chloroacetonacetic acid nethyl ester, pubchem11042 | |
methyl 4-chloro-3-oxobutanoate |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H314
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 251-230-5
RTECSNumber : AK5115000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only