missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-chloro-3-methylbenzoate, 97%

$81.68 - $279.74

Chemical Identifiers

CAS 91367-05-4
Molecular Formula C9H9ClO2
Formula Weight 184.62
MDL Number MFCD00151838
Synonym 4-Chloro-3-methylbenzoic acid methyl ester; 4-Chloro-m-toluic acid methyl ester
Chemical Name or Material Methyl 4-chloro-3-methylbenzoate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1311009 SDS Alfa Aesar™
L1311009
10g Each for $279.74
AAL1311004 SDS Alfa Aesar™
L1311004
2g Each for $81.68
Specifications Description & Specifications

Chemical Identifiers

91367-05-4
184.62
4-Chloro-3-methylbenzoic acid methyl ester; 4-Chloro-m-toluic acid methyl ester
C9H9ClO2
MFCD00151838
Methyl 4-chloro-3-methylbenzoate

Specifications

91367-05-4
184.62
4-Chloro-3-methylbenzoic acid methyl ester; 4-Chloro-m-toluic acid methyl ester
Methyl 4-chloro-3-methylbenzoate
C9H9ClO2
MFCD00151838
10g
97%