missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-bromopyridine-2-carboxylate, ≥97%

Manufacturer:  Alfa Aesar™ H6432714

 View more versions of this product

Catalog No. AAH6432714



Description

Description

Applications
It finds its application as a useful synthetic intermediate and in the straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals. It is used in the synthesis of biaryl mannoside FimH inhibitors for use in urinary infection treatment.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications

Specifications

Methyl 4-bromopyridine-2-carboxylate
25g
C7H6BrNO2
473047
Slightly soluble in water.
COC(=O)C1=NC=CC(=C1)Br
216.034
216.03
≥97%
54°C to 56°C
29681-42-3
MFCD07374897
2-pyridinecarboxylic acid, 4-bromo-, methyl ester, 4-bromo-pyridine-2-carboxylic acid methyl ester, 4-bromopyridine-2-carboxylic acid methyl ester, abbypharma ap-10-5912, acmc-209hag, methyl 4-bromo-2-pyridinecarboxylate, methyl 4-bromo-pyridine-2-carboxylate, methyl 4-bromopicolinate, methyl4-bromopicolinate, pubchem12733
JZFLATQBIPILFS-UHFFFAOYSA-N
methyl 4-bromopyridine-2-carboxylate
12828947
Solid

Chemical Identifiers

29681-42-3
216.034
JZFLATQBIPILFS-UHFFFAOYSA-N
12828947
COC(=O)C1=NC=CC(=C1)Br
C7H6BrNO2
MFCD07374897
2-pyridinecarboxylic acid, 4-bromo-, methyl ester, 4-bromo-pyridine-2-carboxylic acid methyl ester, 4-bromopyridine-2-carboxylic acid methyl ester, abbypharma ap-10-5912, acmc-209hag, methyl 4-bromo-2-pyridinecarboxylate, methyl 4-bromo-pyridine-2-carboxylate, methyl 4-bromopicolinate, methyl4-bromopicolinate, pubchem12733
methyl 4-bromopyridine-2-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

Documents