missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-bromopyridine-2-carboxylate, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6432703

 View more versions of this product

Catalog No. AAH6432703



Description

Description

It finds its application as a useful synthetic intermediate and in the straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals. It is used in the synthesis of biaryl mannoside FimH inhibitors for use in urinary infection treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a useful synthetic intermediate and in the straightforward and efficient synthesis of 3-benzyloxy-4-bromopicolinate ester and 3-benzyloxy-5-bromopicolinate ester, common building blocks for pharmaceuticals and agrochemicals. It is used in the synthesis of biaryl mannoside FimH inhibitors for use in urinary infection treatment.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Specifications

Specifications

Methyl 4-bromopyridine-2-carboxylate
54°C to 56°C
MFCD07374897
473047
Slightly soluble in water.
COC(=O)C1=NC=CC(=C1)Br
216.034
216.03
Solid
29681-42-3
C7H6BrNO2
1 g
methyl 4-bromopicolinate, 4-bromo-pyridine-2-carboxylic acid methyl ester, methyl 4-bromo-2-pyridinecarboxylate, methyl4-bromopicolinate, 2-pyridinecarboxylic acid, 4-bromo-, methyl ester, 4-bromopyridine-2-carboxylic acid methyl ester, abbypharma ap-10-5912, pubchem12733, acmc-209hag, methyl 4-bromo-pyridine-2-carboxylate
JZFLATQBIPILFS-UHFFFAOYSA-N
methyl 4-bromopyridine-2-carboxylate
12828947
≥97%

Chemical Identifiers

29681-42-3
216.034
JZFLATQBIPILFS-UHFFFAOYSA-N
methyl 4-bromopyridine-2-carboxylate
C7H6BrNO2
MFCD07374897
12828947
COC(=O)C1=NC=CC(=C1)Br
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only