Learn More
Methyl 4-bromocrotonate, tech. 85%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1558230
Description
It is a useful synthetic intermediate. It was used in the synthesis of irreversible inhibitors of EGFR and HER-2 tyrosine kinases with enhanced antitumor activities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a useful synthetic intermediate. It was used in the synthesis of irreversible inhibitors of EGFR and HER-2 tyrosine kinases with enhanced antitumor activities.
Solubility
Slightly soluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Methyl 4-bromocrotonate | |
1.524 | |
91°C (195°F) | |
1.501 | |
MFCD00000246,MFCD09998308 | |
UN1760 | |
methyl 4-bromocrotonate, e-methyl 4-bromobut-2-enoate, 4-bromocrotonic acid methyl ester, methyl gamma-bromocrotonate, methyl bromocrotonate, methyl 4-bromo-2-butenoate, 2-butenoic acid, 4-bromo-, methyl ester, methyl 4-bromobut-2-enoate, methyl 2e-4-bromobut-2-enoate, crotonic acid, 4-bromo-, methyl ester | |
RWIKCBHOVNDESJ-NSCUHMNNSA-N | |
methyl (E)-4-bromobut-2-enoate | |
5369175 | |
85% |
1117-71-1 | |
83°C to 85°C (13 mmHg) | |
C5H7BrO2 | |
BrCH2CH=CHCO2CH3 | |
250 g | |
1745755 | |
Slightly soluble in water. | |
COC(=O)\C=C\CBr | |
179.01 | |
179.01 | |
Technical |
Chemical Identifiers
1117-71-1 | |
179.01 | |
RWIKCBHOVNDESJ-NSCUHMNNSA-N | |
5369175 | |
COC(=O)\C=C\CBr |
C5H7BrO2 | |
MFCD00000246,MFCD09998308 | |
methyl 4-bromocrotonate, e-methyl 4-bromobut-2-enoate, 4-bromocrotonic acid methyl ester, methyl gamma-bromocrotonate, methyl bromocrotonate, methyl 4-bromo-2-butenoate, 2-butenoic acid, 4-bromo-, methyl ester, methyl 4-bromobut-2-enoate, methyl 2e-4-bromobut-2-enoate, crotonic acid, 4-bromo-, methyl ester | |
methyl (E)-4-bromobut-2-enoate |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber : 214-251-0
RTECSNumber : GQ3120000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only