Please login to your online account to display your discounted pricing

Methyl 4-bromobutyrate, 98+%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC365830100

 View more versions of this product

Catalog No. AC365830100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Liquid
Formula Weight 181.03
Quantity 10g
Color Clear colorless to pale yellow
Density 1.4340g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 Warning
Molecular Formula C5H9BrO2
Assay Percent Range 98+%
Synonym 4-Bromobutyric acid methyl ester
Refractive Index 1.4610 to 1.4630 (20°C, 589nm)
Chemical Name or Material Methyl 4-bromobutyrate
CAS 4897-84-1
MDL Number MFCD00041482
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

Specifications

Chemical Name or Material Methyl 4-bromobutyrate
CAS 4897-84-1
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C5H9BrO2
Formula Weight 181.03
MDL Number MFCD00041482
Synonym 4-Bromobutyric acid methyl ester
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents