missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-bromobutyrate, 98+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 365830500

 View more versions of this product

Catalog No. AC365830500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Methyl 4-bromobutyrate
1.4340g/mL
87°C
98% min. (GC)
C5H9BrO2
MFCD00041482
1.434
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
COC(=O)CCCBr
181.03
181.03
4897-84-1
186.0°C to 187.0°C
Authentic
Glass bottle
1.4610 to 1.4630 (20°C, 589nm)
50g
methyl 4-bromobutyrate, 4-bromobutyric acid methyl ester, methyl-4-bromobutyrate, butanoic acid, 4-bromo-, methyl ester, methyl4-bromobutyrate, butyric acid, 4-bromo-, methyl ester, methyl gamma-bromobutyrate, acmc-1amn9, 4-brombutyric acid methyl ester, 4-bromobutyric acid methyl-ester
QAWFLJGZSZIZHO-UHFFFAOYSA-N
methyl 4-bromobutanoate
107604
98+%

Chemical Identifiers

4897-84-1
181.03
QAWFLJGZSZIZHO-UHFFFAOYSA-N
107604
COC(=O)CCCBr
C5H9BrO2
MFCD00041482
methyl 4-bromobutyrate, 4-bromobutyric acid methyl ester, methyl-4-bromobutyrate, butanoic acid, 4-bromo-, methyl ester, methyl4-bromobutyrate, butyric acid, 4-bromo-, methyl ester, methyl gamma-bromobutyrate, acmc-1amn9, 4-brombutyric acid methyl ester, 4-bromobutyric acid methyl-ester
methyl 4-bromobutanoate
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Call a POISON CENTER or doctor/physicia

GHS Signal Word: Warning

EINECSNumber : 225-523-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only