Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4'-bromobiphenyl-4-carboxylate, 95%

$74.42 - $390.78

Chemical Identifiers

Chemical Name or Material Methyl 4'-bromobiphenyl-4-carboxylate
CAS 89901-03-1
Molecular Formula C14H11BrO2
Formula Weight 291.14g/mol
MDL Number MFCD00496638
Synonym 4'-Bromobiphenyl-4-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5192003

 
alfa aesar™
H51920-03
Each for $74.42

AAH5192006

 
alfa aesar™
H51920-06
Each for $390.78
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material Methyl 4'-bromobiphenyl-4-carboxylate
CAS 89901-03-1
Assay 95%
Molecular Formula C14H11BrO2
Formula Weight 291.14g/mol
MDL Number MFCD00496638
Synonym 4'-Bromobiphenyl-4-carboxylic acid methyl ester

Applications
It is employed in the design, synthesis, and biological evaluation of novel nonsteroidal farnesoid x receptor (fxr) antagonists.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.