Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4'-bromobiphenyl-3-carboxylate, 95%

$62.34 - $250.91

Chemical Identifiers

Chemical Name or Material Methyl 4'-bromobiphenyl-3-carboxylate
CAS 149506-25-2
Molecular Formula C14H11BrO2
Formula Weight 291.14
MDL Number MFCD05980426
Synonym 4'-Bromobiphenyl-3-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5190503

  SDS
alfa aesar™
H5190503
1g Each for $62.34

AAH5190506

  SDS
alfa aesar™
H5190506
5g Each for $250.91
Description & Specifications

Specifications

Formula Weight 291.14
Chemical Name or Material Methyl 4'-bromobiphenyl-3-carboxylate
Assay 95%
Molecular Formula C14H11BrO2
MDL Number MFCD05980426
CAS 149506-25-2
Synonym 4'-Bromobiphenyl-3-carboxylic acid methyl ester
Quantity 1g