missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-bromobenzoate, 98+%

Manufacturer:  Alfa Aesar™ A1551722

 View more versions of this product

Catalog No. AAA1551722



Description

Description

Applications
Methyl 4-bromobenzoate is used as a starting material in the synthesis of three-carbon-bridged 5-substituted furo[2,3-d]pyrimidine and 6-substituted pyrrolo[2,3-d]pyrimidine and methyl 4-tri-n-butylstannylbenzoate. It is also used to stimulate microbial dechlorination of polychlorinated biphenyls. Further, it is used in the preparation of stable radioiodinating reagent to label monoclonal antibodies for radiotherapy of cancer.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Methyl 4-bromobenzoate
100g
C8H7BrO2
2045132
benzoic acid, 4-bromo-, methyl ester, methyl p-bromobenzoate, 4-bromobenzoic acid methyl ester, p-bromobenzoic acid, methyl ester, unii-ub1ysj5xar, ub1ysj5xar, benzoic acid, p-bromo-, methyl ester, pubchem3962, methyl-4-bromobenzoate, methyl p-bromo benzoate
COC(=O)C1=CC=C(C=C1)Br
215.046
215.05
77°C to 81°C
619-42-1
MFCD00013531
14,1408
CZNGTXVOZOWWKM-UHFFFAOYSA-N
methyl 4-bromobenzoate
12081
≥98%

Chemical Identifiers

619-42-1
215.046
CZNGTXVOZOWWKM-UHFFFAOYSA-N
12081
COC(=O)C1=CC=C(C=C1)Br
C8H7BrO2
MFCD00013531
benzoic acid, 4-bromo-, methyl ester, methyl p-bromobenzoate, 4-bromobenzoic acid methyl ester, p-bromobenzoic acid, methyl ester, unii-ub1ysj5xar, ub1ysj5xar, benzoic acid, p-bromo-, methyl ester, pubchem3962, methyl-4-bromobenzoate, methyl p-bromo benzoate
methyl 4-bromobenzoate
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents