missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 4-Bromo-3-methylbenzoate 97.0+%, TCI America™
Supplier: TCI America B225625G
Specifications
Methyl 4-Bromo-3-methylbenzoate | |
43°C | |
C9H9BrO2 | |
25 g | |
GTZTYNPAPQKIIR-UHFFFAOYSA-N | |
methyl 4-bromo-3-methylbenzoate | |
3294148 | |
≥97.0% (GC) |
148547-19-7 | |
White-Yellow | |
MFCD00673014 | |
4-bromo-3-methylbenzoic acid methyl ester, methyl4-bromo-3-methylbenzoate, methyl 4-bromo-m-toluate, 4-bromo-3-methyl-benzoic acid methyl ester, methyl 4-bromo-3-methyl-benzoate, 4-bromo-m-toluic acid methyl ester, benzoic acid, 4-bromo-3-methyl-, methyl ester, 2-bromo-5-methoxycarbonyl toluene, 3-methyl-4-bromobenzoic acid methyl ester, pubchem3970 | |
CC1=C(C=CC(=C1)C(=O)OC)Br | |
229.073 | |
229.07 | |
Crystalline Powder |
Chemical Identifiers
148547-19-7 | |
229.073 | |
GTZTYNPAPQKIIR-UHFFFAOYSA-N | |
3294148 | |
CC1=C(C=CC(=C1)C(=O)OC)Br |
C9H9BrO2 | |
MFCD00673014 | |
4-bromo-3-methylbenzoic acid methyl ester, methyl4-bromo-3-methylbenzoate, methyl 4-bromo-m-toluate, 4-bromo-3-methyl-benzoic acid methyl ester, methyl 4-bromo-3-methyl-benzoate, 4-bromo-m-toluic acid methyl ester, benzoic acid, 4-bromo-3-methyl-, methyl ester, 2-bromo-5-methoxycarbonyl toluene, 3-methyl-4-bromobenzoic acid methyl ester, pubchem3970 | |
methyl 4-bromo-3-methylbenzoate |
Safety and Handling
TSCA : No