missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 4-bromo-2-fluorophenylacetate, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H62482MD
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl 4-bromo-2-fluorophenylacetate | |
C9H8BrFO2 | |
250 mg | |
QBGQCRUYARHQGO-UHFFFAOYSA-N | |
methyl 2-(4-bromo-2-fluorophenyl)acetate | |
53429406 | |
96% |
193290-19-6 | |
MFCD20483545 | |
methyl 2-4-bromo-2-fluorophenyl acetate, methyl 4-bromo-2-fluorophenylacetate, methyl 4-bromo-2-fluorophenyl acetate, methyl 2-4-bromo-2-fluoro-phenyl acetate, 4-bromo-2-fluorophenyl acetic acid methyl ester, 4-bromo-2-fluoro-phenyl-acetic acid methyl ester, benzeneacetic acid, 4-bromo-2-fluoro-, methyl ester | |
COC(=O)CC1=C(F)C=C(Br)C=C1 | |
247.06 | |
247.06 | |
Solid |
Chemical Identifiers
193290-19-6 | |
247.06 | |
QBGQCRUYARHQGO-UHFFFAOYSA-N | |
53429406 | |
COC(=O)CC1=C(F)C=C(Br)C=C1 |
C9H8BrFO2 | |
MFCD20483545 | |
methyl 2-4-bromo-2-fluorophenyl acetate, methyl 4-bromo-2-fluorophenylacetate, methyl 4-bromo-2-fluorophenyl acetate, methyl 2-4-bromo-2-fluoro-phenyl acetate, 4-bromo-2-fluorophenyl acetic acid methyl ester, 4-bromo-2-fluoro-phenyl-acetic acid methyl ester, benzeneacetic acid, 4-bromo-2-fluoro-, methyl ester | |
methyl 2-(4-bromo-2-fluorophenyl)acetate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only