missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-Bromo-2-(bromomethyl)benzoate, TCI America™

$55.00 - $213.89

Chemical Identifiers

CAS 78471-43-9
Molecular Formula C9H8Br2O2
Molecular Weight (g/mol) 307.969
MDL Number MFCD04114319
InChI Key SGFACFBLUAWICV-UHFFFAOYSA-N
Synonym methyl 4-bromo-2-bromomethyl benzoate, methyl 4-bromo-2-bromomethyl-benzoate, 4-bromo-2-bromomethylbenzoic acid methyl ester, 4-bromo-2-bromomethyl-benzoic acid methyl ester, benzoic acid, 4-bromo-2-bromomethyl-, methyl ester, 4-bromo-2-bromomethyl benzoic acid methyl ester, acmc-209pec, methyl alpha,5-dibromo-o-toluate, methyl 4bromo-2-bromomethylbenzoate, methyl4-bromo-2-bromomethylbenzoate
PubChem CID 22031079
IUPAC Name methyl 4-bromo-2-(bromomethyl)benzoate
SMILES COC(=O)C1=C(C=C(C=C1)Br)CBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M29411G View Documents TCI America
M29411G
1g Each for $213.89
 
M2941200MG View Documents TCI America
M2941200MG
200mg Each for $55.00
 
Specifications

Chemical Identifiers

78471-43-9
307.969
SGFACFBLUAWICV-UHFFFAOYSA-N
22031079
COC(=O)C1=C(C=C(C=C1)Br)CBr
C9H8Br2O2
MFCD04114319
methyl 4-bromo-2-bromomethyl benzoate, methyl 4-bromo-2-bromomethyl-benzoate, 4-bromo-2-bromomethylbenzoic acid methyl ester, 4-bromo-2-bromomethyl-benzoic acid methyl ester, benzoic acid, 4-bromo-2-bromomethyl-, methyl ester, 4-bromo-2-bromomethyl benzoic acid methyl ester, acmc-209pec, methyl alpha,5-dibromo-o-toluate, methyl 4bromo-2-bromomethylbenzoate, methyl4-bromo-2-bromomethylbenzoate
methyl 4-bromo-2-(bromomethyl)benzoate

Specifications

Undesignated
1g
C9H8Br2O2
3261
SGFACFBLUAWICV-UHFFFAOYSA-N
methyl 4-bromo-2-(bromomethyl)benzoate
22031079
Crystal-Powder at 20°C
Methyl 4-Bromo-2-(bromomethyl)benzoate
81°C
78471-43-9
MFCD04114319
methyl 4-bromo-2-bromomethyl benzoate, methyl 4-bromo-2-bromomethyl-benzoate, 4-bromo-2-bromomethylbenzoic acid methyl ester, 4-bromo-2-bromomethyl-benzoic acid methyl ester, benzoic acid, 4-bromo-2-bromomethyl-, methyl ester, 4-bromo-2-bromomethyl benzoic acid methyl ester, acmc-209pec, methyl alpha,5-dibromo-o-toluate, methyl 4bromo-2-bromomethylbenzoate, methyl4-bromo-2-bromomethylbenzoate
COC(=O)C1=C(C=C(C=C1)Br)CBr
307.969
307.97
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator