missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-Bromo-2-(bromomethyl)benzoate 97.0+%, TCI America™

Manufacturer:  TCI America M29411G

 View more versions of this product

Catalog No. M29411G



Specifications

Specifications

Methyl 4-Bromo-2-(bromomethyl)benzoate
81°C
78471-43-9
MFCD04114319
methyl 4-bromo-2-bromomethyl benzoate, methyl 4-bromo-2-bromomethyl-benzoate, 4-bromo-2-bromomethylbenzoic acid methyl ester, 4-bromo-2-bromomethyl-benzoic acid methyl ester, benzoic acid, 4-bromo-2-bromomethyl-, methyl ester, 4-bromo-2-bromomethyl benzoic acid methyl ester, acmc-209pec, methyl alpha,5-dibromo-o-toluate, methyl 4bromo-2-bromomethylbenzoate, methyl4-bromo-2-bromomethylbenzoate
COC(=O)C1=C(C=C(C=C1)Br)CBr
307.969
307.97
≥97.0% (GC)
Undesignated
1g
C9H8Br2O2
3261
SGFACFBLUAWICV-UHFFFAOYSA-N
methyl 4-bromo-2-(bromomethyl)benzoate
22031079
Crystal-Powder at 20°C

Chemical Identifiers

78471-43-9
307.969
SGFACFBLUAWICV-UHFFFAOYSA-N
22031079
COC(=O)C1=C(C=C(C=C1)Br)CBr
C9H8Br2O2
MFCD04114319
methyl 4-bromo-2-bromomethyl benzoate, methyl 4-bromo-2-bromomethyl-benzoate, 4-bromo-2-bromomethylbenzoic acid methyl ester, 4-bromo-2-bromomethyl-benzoic acid methyl ester, benzoic acid, 4-bromo-2-bromomethyl-, methyl ester, 4-bromo-2-bromomethyl benzoic acid methyl ester, acmc-209pec, methyl alpha,5-dibromo-o-toluate, methyl 4bromo-2-bromomethylbenzoate, methyl4-bromo-2-bromomethylbenzoate
methyl 4-bromo-2-(bromomethyl)benzoate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents