Learn More
Methyl (+)-4-Boc-piperazine-2-carboxylate, Thermo Scientific Chemicals
$281.23 - $848.77
Chemical Identifiers
CAS | 129799-08-2 |
---|---|
Molecular Formula | C11H21N2O4 |
Molecular Weight (g/mol) | 245.30 |
MDL Number | MFCD01632465,MFCD04115325,MFCD04115326 |
InChI Key | QUKAHFCVKNRRBU-QMMMGPOBSA-O |
Synonym | methyl 4-boc-piperazine-2-carboxylate, 1-tert-butyl 3-methyl piperazine-1,3-dicarboxylate, n-4-boc-2-piperazinecarboxylic acid methyl ester, 1-n-boc-piperazine-3-carboxylic acid methyl ester, 4n-boc-piperazine-2-carboxylic acid methyl ester, piperazine-1,3-dicarboxylic acid 1-tert-butyl ester 3-methyl ester, 1,3-piperazinedicarboxylic acid, 1-1,1-dimethylethyl 3-methyl ester, 1-n-boc-piperazine-3-carboxylic acid methyl ester; piperazine-1,3-dicarboxylic acid 1-tert-butyl ester 3-methyl ester, tert-butyl 3-methoxycarbonyl piperazinecarboxylate, pubchem18338 |
PubChem CID | 2756819 |
IUPAC Name | 1-O-tert-butyl 3-O-methyl piperazine-1,3-dicarboxylate |
SMILES | COC(=O)[C@@H]1CN(CC[NH2+]1)C(=O)OC(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH2788603
|
Thermo Scientific Chemicals
H2788603 |
1 g |
Each for $281.23
|
|
|||||
AAH2788606
|
Thermo Scientific Chemicals
H2788606 |
5 g |
Each for $848.77
|
|
|||||
Description
It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as pharmaceutical intermediate.
Solubility
Soluble in dimethyl sulfoxide.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Chemical Identifiers
129799-08-2 | |
245.30 | |
QUKAHFCVKNRRBU-QMMMGPOBSA-O | |
2756819 | |
COC(=O)[C@@H]1CN(CC[NH2+]1)C(=O)OC(C)(C)C |
C11H21N2O4 | |
MFCD01632465,MFCD04115325,MFCD04115326 | |
methyl 4-boc-piperazine-2-carboxylate, 1-tert-butyl 3-methyl piperazine-1,3-dicarboxylate, n-4-boc-2-piperazinecarboxylic acid methyl ester, 1-n-boc-piperazine-3-carboxylic acid methyl ester, 4n-boc-piperazine-2-carboxylic acid methyl ester, piperazine-1,3-dicarboxylic acid 1-tert-butyl ester 3-methyl ester, 1,3-piperazinedicarboxylic acid, 1-1,1-dimethylethyl 3-methyl ester, 1-n-boc-piperazine-3-carboxylic acid methyl ester; piperazine-1,3-dicarboxylic acid 1-tert-butyl ester 3-methyl ester, tert-butyl 3-methoxycarbonyl piperazinecarboxylate, pubchem18338 | |
1-O-tert-butyl 3-O-methyl piperazine-1,3-dicarboxylate |
Specifications
129799-08-2 | |
MFCD01632465,MFCD04115325,MFCD04115326 | |
methyl 4-boc-piperazine-2-carboxylate, 1-tert-butyl 3-methyl piperazine-1,3-dicarboxylate, n-4-boc-2-piperazinecarboxylic acid methyl ester, 1-n-boc-piperazine-3-carboxylic acid methyl ester, 4n-boc-piperazine-2-carboxylic acid methyl ester, piperazine-1,3-dicarboxylic acid 1-tert-butyl ester 3-methyl ester, 1,3-piperazinedicarboxylic acid, 1-1,1-dimethylethyl 3-methyl ester, 1-n-boc-piperazine-3-carboxylic acid methyl ester; piperazine-1,3-dicarboxylic acid 1-tert-butyl ester 3-methyl ester, tert-butyl 3-methoxycarbonyl piperazinecarboxylate, pubchem18338 | |
QUKAHFCVKNRRBU-QMMMGPOBSA-O | |
1-O-tert-butyl 3-O-methyl piperazine-1,3-dicarboxylate | |
2756819 | |
Methyl (+/-)-4-Boc-piperazine-2-carboxylate |
C11H21N2O4 | |
1 g | |
Soluble in dimethyl sulfoxide. | |
COC(=O)[C@@H]1CN(CC[NH2+]1)C(=O)OC(C)(C)C | |
245.30 | |
244.29 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only