missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-Benzyloxybenzoate, TCI America™

$230.70 - $786.83

Chemical Identifiers

CAS 32122-11-5
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.274
MDL Number MFCD00017613
InChI Key ZLLQTDQOTFCCDF-UHFFFAOYSA-N
Synonym methyl 4-benzyloxybenzoate, methyl 4-benzyloxy benzoate, 4-benzyloxybenzoic acid methyl ester, methyl4-benzyloxybenzoate, methyl 4-benzyloxy benzenecarboxylate, methyl 4-phenylmethoxy benzoate, methyl-4-benzyloxybenzoate, acmc-209hr7, 4-benzyloxy-benzoic acid methyl ester, benzoic acid,4-phenylmethoxy-, methyl ester
PubChem CID 299810
IUPAC Name methyl 4-phenylmethoxybenzoate
SMILES COC(=O)C1=CC=C(C=C1)OCC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M218025G View Documents TCI America
M218025G
25g Each for $786.83
 
M21805G View Documents TCI America
M21805G
5g Each for $230.70
 
Specifications

Chemical Identifiers

32122-11-5
242.274
ZLLQTDQOTFCCDF-UHFFFAOYSA-N
299810
COC(=O)C1=CC=C(C=C1)OCC2=CC=CC=C2
C15H14O3
MFCD00017613
methyl 4-benzyloxybenzoate, methyl 4-benzyloxy benzoate, 4-benzyloxybenzoic acid methyl ester, methyl4-benzyloxybenzoate, methyl 4-benzyloxy benzenecarboxylate, methyl 4-phenylmethoxy benzoate, methyl-4-benzyloxybenzoate, acmc-209hr7, 4-benzyloxy-benzoic acid methyl ester, benzoic acid,4-phenylmethoxy-, methyl ester
methyl 4-phenylmethoxybenzoate

Specifications

White
25g
C15H14O3
methyl 4-benzyloxybenzoate, methyl 4-benzyloxy benzoate, 4-benzyloxybenzoic acid methyl ester, methyl4-benzyloxybenzoate, methyl 4-benzyloxy benzenecarboxylate, methyl 4-phenylmethoxy benzoate, methyl-4-benzyloxybenzoate, acmc-209hr7, 4-benzyloxy-benzoic acid methyl ester, benzoic acid,4-phenylmethoxy-, methyl ester
COC(=O)C1=CC=C(C=C1)OCC2=CC=CC=C2
242.274
242.27
≥98.0% (GC)
99°C
32122-11-5
MFCD00017613
ZLLQTDQOTFCCDF-UHFFFAOYSA-N
methyl 4-phenylmethoxybenzoate
299810
Crystal-Powder at 20°C
Methyl 4-Benzyloxybenzoate
Safety and Handling

Safety and Handling

TSCA : No