Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-benzyloxybenzoate, 97%

$84.97 - $315.55

Chemical Identifiers

Chemical Name or Material Methyl 4-benzyloxybenzoate
CAS 32122-11-5
Molecular Formula C15H14O3
Formula Weight 242.27
MDL Number MFCD00017613
Beilstein 2119212
Synonym 4-Benzyloxybenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0874906

 
alfa aesar™
L0874906
5g Each for $84.97

AAL0874914

 
alfa aesar™
L0874914
25g Each for $315.55
Description & Specifications

Specifications

Formula Weight 242.27
Chemical Name or Material Methyl 4-benzyloxybenzoate
Molecular Formula C15H14O3
MDL Number MFCD00017613
CAS 32122-11-5
Beilstein 2119212
Synonym 4-Benzyloxybenzoic acid methyl ester
Quantity 5g
Assay 97%
Melting Point 96° to 100°C