missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-aminosalicylate, 97%

Manufacturer:  Alfa Aesar™ 4552022

 View more versions of this product

Catalog No. AA4552022



Specifications

Specifications

Methyl 4-aminosalicylate
4136-97-4
MFCD00088091
QQOXBFUTRLDXDP-UHFFFAOYSA-N
methyl 4-amino-2-hydroxybenzoate
77787
167.18
97%
100g
C8H9NO3
methyl 4-aminosalicylate, methyl p-aminosalicylate, pas methyl ester, p-aminosalicylic acid methyl ester, methyl 4-amino-2-hydroxybenzenecarboxylate, benzoic acid, 4-amino-2-hydroxy-, methyl ester, methyl 4-amino-2-hydroxy-benzoate, salicylic acid, amino-, methyl ester, 4-aminosalicylic acid methyl ester, 4-amino-2-hydroxy-benzoic acid methyl ester
COC(=O)C1=C(C=C(C=C1)N)O
167.164
CHEBI:35089
Powder

Chemical Identifiers

4136-97-4
167.164
QQOXBFUTRLDXDP-UHFFFAOYSA-N
77787
methyl 4-amino-2-hydroxybenzoate
C8H9NO3
MFCD00088091
methyl 4-aminosalicylate, methyl p-aminosalicylate, pas methyl ester, p-aminosalicylic acid methyl ester, methyl 4-amino-2-hydroxybenzenecarboxylate, benzoic acid, 4-amino-2-hydroxy-, methyl ester, methyl 4-amino-2-hydroxy-benzoate, salicylic acid, amino-, methyl ester, 4-aminosalicylic acid methyl ester, 4-amino-2-hydroxy-benzoic acid methyl ester
CHEBI:35089
COC(=O)C1=C(C=C(C=C1)N)O
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents