Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-amino-3-hydroxybenzoate, 98%


Quantity 10g

Manufacturer: alfa aesar™  L0568709

 View more versions of this product

Catalog No. AAL0568709

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 167.17
Chemical Name or Material Methyl 4-amino-3-hydroxybenzoate
Assay 98%
CAS 63435-16-5
Molecular Formula C8H9NO3
MDL Number MFCD00017093
Beilstein 608716
Synonym 4-Amino-3-hydroxybenzoic acid methyl ester
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Melting Point 118° to 122°C
Quantity 10g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Specifications

Chemical Name or Material Methyl 4-amino-3-hydroxybenzoate
CAS 63435-16-5
Molecular Formula C8H9NO3
Formula Weight 167.17
MDL Number MFCD00017093
Beilstein 608716
Synonym 4-Amino-3-hydroxybenzoic acid methyl ester

Applications
Methyl 4-amino-3-hydroxybenzoate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Notes
Incompatible with Strong oxidizing agents, Strong acids and bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Accessories

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.