Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-amino-3-hydroxybenzoate, 98%

$123.55 - $474.64

Chemical Identifiers

Chemical Name or Material Methyl 4-amino-3-hydroxybenzoate
CAS 63435-16-5
Molecular Formula C8H9NO3
Formula Weight 167.17
MDL Number MFCD00017093
Beilstein 608716
Synonym 4-Amino-3-hydroxybenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0568704

  SDS
alfa aesar™
L0568704
2g Each for $123.55

AAL0568709

  SDS
alfa aesar™
L0568709
10g Each for $474.64
Description & Specifications

Specifications

Formula Weight 167.17
CAS 63435-16-5
Assay 98%
Synonym 4-Amino-3-hydroxybenzoic acid methyl ester
Chemical Name or Material Methyl 4-amino-3-hydroxybenzoate
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 118° to 122°C
MDL Number MFCD00017093
Health Hazard 1 Warning
Molecular Formula C8H9NO3
Quantity 2g
Beilstein 608716

Applications
Methyl 4-amino-3-hydroxybenzoate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Notes
Incompatible with Strong oxidizing agents, Strong acids and bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.