missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 4-amino-3-chlorobenzoate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2251906
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl 4-amino-3-chlorobenzoate | |
112°C to 113°C | |
MFCD01320686 | |
4-amino-3-chlorobenzoic acid methyl ester, 4-amino-3-chloro-benzoic acid methyl ester, methyl 4-amino-3-chloro-benzoate, benzoic acid, 4-amino-3-chloro-, methyl ester, 2-chloro-4-methoxycarbonyl aniline, pubchem10804, akos bb-3098, timtec-bb sbb003712, acmc-20a51m, methyl-4-amino-3-chlorobenzoate | |
COC(=O)C1=CC(=C(C=C1)N)Cl | |
185.607 | |
185.61 |
84228-44-4 | |
C8H8ClNO2 | |
5 g | |
WKVJBYKFGNVWLM-UHFFFAOYSA-N | |
methyl 4-amino-3-chlorobenzoate | |
1515284 | |
99% |
Chemical Identifiers
84228-44-4 | |
185.607 | |
WKVJBYKFGNVWLM-UHFFFAOYSA-N | |
1515284 | |
COC(=O)C1=CC(=C(C=C1)N)Cl |
C8H8ClNO2 | |
MFCD01320686 | |
4-amino-3-chlorobenzoic acid methyl ester, 4-amino-3-chloro-benzoic acid methyl ester, methyl 4-amino-3-chloro-benzoate, benzoic acid, 4-amino-3-chloro-, methyl ester, 2-chloro-4-methoxycarbonyl aniline, pubchem10804, akos bb-3098, timtec-bb sbb003712, acmc-20a51m, methyl-4-amino-3-chlorobenzoate | |
methyl 4-amino-3-chlorobenzoate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only