missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 4-Acetylbenzoate, TCI America™

$42.78 - $99.31

Chemical Identifiers

CAS 3609-53-8
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD00216474
InChI Key QNTSFZXGLAHYLC-UHFFFAOYSA-N
Synonym benzoic acid, 4-acetyl-, methyl ester, unii-3s3d53j4nt, 4-acetylbenzoic acid methyl ester, methyl4-acetylbenzoate, 4-acetyl-benzoic acid methyl ester, methyl 4-acetyl benzoate, pubchem22273, acmc-209ikx, #, ksc497k9b
PubChem CID 137990
IUPAC Name methyl 4-acetylbenzoate
SMILES CC(=O)C1=CC=C(C=C1)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M21635G View Documents TCI America
M21635G
5g Each for $42.78
 
M216325G View Documents TCI America
M216325G
25g Each for $99.31
 
Specifications

Chemical Identifiers

3609-53-8
178.187
QNTSFZXGLAHYLC-UHFFFAOYSA-N
137990
CC(=O)C1=CC=C(C=C1)C(=O)OC
C10H10O3
MFCD00216474
benzoic acid, 4-acetyl-, methyl ester, unii-3s3d53j4nt, 4-acetylbenzoic acid methyl ester, methyl4-acetylbenzoate, 4-acetyl-benzoic acid methyl ester, methyl 4-acetyl benzoate, pubchem22273, acmc-209ikx, #, ksc497k9b
methyl 4-acetylbenzoate

Specifications

White-Yellow
5g
C10H10O3
benzoic acid, 4-acetyl-, methyl ester, unii-3s3d53j4nt, 4-acetylbenzoic acid methyl ester, methyl4-acetylbenzoate, 4-acetyl-benzoic acid methyl ester, methyl 4-acetyl benzoate, pubchem22273, acmc-209ikx, #, ksc497k9b
CC(=O)C1=CC=C(C=C1)C(=O)OC
178.187
178.19
≥98.0% (GC)
97°C
3609-53-8
MFCD00216474
QNTSFZXGLAHYLC-UHFFFAOYSA-N
methyl 4-acetylbenzoate
137990
Crystal-Powder at 20°C
Methyl 4-Acetylbenzoate
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1406

TSCA : No