Please login to your online account to display your discounted pricing

Methyl 4-acetylbenzoate, 99+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC424770050

 View more versions of this product

Catalog No. AC424770050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 178.19
Physical Form Amorphous Powder
Quantity 5g
Color White to pale yellow
Melting Point 93° to 95°C
Infrared Spectrum Authentic
Molecular Formula C10H10O3
Linear Formula CH3OOCC6H4COCH3
Assay Percent Range 99+%
MDL Number MFCD00216474
Packaging Glass bottle
Chemical Name or Material Methyle4-acetylbenzoate
CAS 3609-53-8

Specifications

Chemical Name or Material Methyle4-acetylbenzoate
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C10H10O3
Linear Formula CH3OOCC6H4COCH3
Formula Weight 178.19
MDL Number MFCD00216474