Please login to your online account to display your discounted pricing

Methyl 4-acetylbenzoate, 99+%, ACROS Organics™

$42.36 - $42.36

Chemical Identifiers

Chemical Name or Material Methyle4-acetylbenzoate
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C10H10O3
Linear Formula CH3OOCC6H4COCH3
Formula Weight 178.19
MDL Number MFCD00216474
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC424770050

  SDS
thermo fisher scientific
AC424770050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC424770010

  SDS
thermo fisher scientific
424770010
Glass bottle 1g Each for $42.36
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Amorphous Powder
Formula Weight 178.19
Quantity 5g
Color White to pale yellow
Melting Point 93° to 95°C
Infrared Spectrum Authentic
Molecular Formula C10H10O3
Linear Formula CH3OOCC6H4COCH3
Chemical Name or Material Methyle4-acetylbenzoate
Assay Percent Range 99+%
MDL Number MFCD00216474
Packaging Glass bottle
CAS 3609-53-8

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.