Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-acetylbenzoate, 99%

$103.28 - $400.22

Chemical Identifiers

Chemical Name or Material Methyl 4-acetylbenzoate
CAS 3609-53-8
Molecular Formula C10H10O3
Formula Weight 178.19
MDL Number MFCD00216474
Synonym 4-Acetylbenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2162604

 
alfa aesar™
B2162604
2g Each for $103.28

AAB2162609

 
alfa aesar™
B2162609
10g Each for $400.22
Description & Specifications

Specifications

Formula Weight 178.19
Molecular Formula C10H10O3
Melting Point 92° to 95°C
Chemical Name or Material Methyl 4-acetylbenzoate
Assay 99%
MDL Number MFCD00216474
CAS 3609-53-8
Synonym 4-Acetylbenzoic acid methyl ester
Quantity 2g