Please login to your online account to display your discounted pricing

Methyl 4-acetamido-5-chloro-2-methoxybenzoate, +99%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  270610100

 View more versions of this product

Catalog No. AC270610100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Fine Crystalline Powder
Formula Weight 257.66
Quantity 10g
Color White to light beige
Melting Point 153° to 156°C
EINECS Number 223-840-1
TSCA TSCA
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Methyl 4-acetamido-5-chloro-2-methoxybenzoate
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C11H12ClNO4
MDL Number MFCD00048191
CAS 4093-31-6

Specifications

Chemical Name or Material Methyl 4-acetamido-5-chloro-2-methoxybenzoate
Name Note 99+%
CAS 4093-31-6
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C11H12ClNO4
Formula Weight 257.66
MDL Number MFCD00048191