Please login to your online account to display your discounted pricing

Methyl 4-acetamido-5-chloro-2-methoxybenzoate, +99%, ACROS Organics™

$25.99 - $25.99

Chemical Identifiers

Chemical Name or Material Methyl 4-acetamido-5-chloro-2-methoxybenzoate
Name Note 99+%
CAS 4093-31-6
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C11H12ClNO4
Formula Weight 257.66
MDL Number MFCD00048191
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC270610100

  SDS
thermo fisher scientific
270610100
Glass bottle 10g Each for $25.99
Description & Specifications

Specifications

Assay ≥99.0%
Formula Weight 257.66
Physical Form Fine Crystalline Powder
Quantity 10g
Color White to light beige
Melting Point 153° to 156°C
TSCA TSCA
EINECS Number 223-840-1
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99+%
MDL Number MFCD00048191
CAS 4093-31-6
Assay Percent Range 99+%
Molecular Formula C11H12ClNO4
Chemical Name or Material Methyl 4-acetamido-5-chloro-2-methoxybenzoate