Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 4-acetamido-5-chloro-2-methoxybenzoate, 98%

$142.07 - $142.07

Chemical Identifiers

Chemical Name or Material Methyl 4-acetamido-5-chloro-2-methoxybenzoate
CAS 4093-31-6
Molecular Formula C11H12ClNO4
Formula Weight 257.67
MDL Number MFCD00048191
Synonym 4-Acetamido-5-chloro-2-methoxybenzoic acid methyl ester; Methyl 4-acetamido-5-chloro-o-anisate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2086918

 
alfa aesar™
B2086918
50g Each for $142.07
Description & Specifications

Specifications

Formula Weight 257.67
Chemical Name or Material Methyl 4-acetamido-5-chloro-2-methoxybenzoate
TSCA TSCA
Assay 98%
Molecular Formula C11H12ClNO4
MDL Number MFCD00048191
CAS 4093-31-6
Synonym 4-Acetamido-5-chloro-2-methoxybenzoic acid methyl ester; Methyl 4-acetamido-5-chloro-o-anisate
Health Hazard 1 Warning
Melting Point 153° to 156°C
Quantity 50g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.